Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265909.1 | NP_001252838.1 | 230 | 125 | ||
XM_015122786.2 | XP_014978272.1 | 547 | 125 | |||
GenBank mRNA | JU337115.1 | - | 230 | 125 | ||
JV727064.1 | - | 192 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |