Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258151.1 | NP_001245080.1 | 567 | 93 | ||
GenBank mRNA | JU321201.1 | - | 546 | 93 | ||
JU473292.1 | - | 567 | 93 | |||
JV043616.1 | - | 567 | 93 | |||
JV638609.1 | - | 583 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |