Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260694.1 | NP_001247623.1 | 379 | 70 | ||
XM_015132325.2 | XP_014987811.1 | 423 | 70 | |||
GenBank mRNA | JU474171.1 | - | 379 | 70 | ||
JV635932.1 | - | 368 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |