Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194731.2 | NP_001181660.1 | 456 | 115 | ||
GenBank mRNA | JU332828.1 | - | 265 | 115 | ||
JU474239.1 | - | 384 | 115 | |||
JV046132.1 | - | 256 | 115 | |||
JV601388.1 | - | 303 | 115 | |||
JV638766.1 | - | 419 | 115 | |||
JV672360.1 | - | 274 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |