Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104381.4 | XP_001104381.1 | 508 | 133 | ||
XM_015142783.2 | XP_014998269.1 | 505 | 150 | |||
XM_015142784.2 | XP_014998270.1 | 489 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |