Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271646.1 | NP_001258575.1 | 953 | 148 | ||
XM_015113722.2 | XP_014969208.2 | 1406 | 148 | |||
XM_015113724.2 | XP_014969210.2 | 1304 | 148 | |||
XM_028832812.1 | XP_028688645.1 | 696 | 148 | |||
GenBank mRNA | AB738913.1 | - | 953 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |