Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257642.2 | NP_001244571.1 | 451 | 156 | ||
XM_028830388.1 | XP_028686221.1 | 772 | 156 | |||
XM_028830389.1 | XP_028686222.1 | 747 | 156 | |||
GenBank mRNA | JU337033.1 | - | 452 | 156 | ||
JU473225.1 | - | 749 | 156 | |||
JV044588.1 | - | 610 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |