Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265667.1 | NP_001252596.1 | 1226 | 77 | ||
XM_028832850.1 | XP_028688683.1 | 1370 | 77 | |||
GenBank mRNA | JV600168.1 | - | 1226 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |