Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258112.1 | NP_001245041.1 | 948 | 59 | ||
GenBank mRNA | JU322144.1 | - | 953 | 59 | ||
JU477886.1 | - | 948 | 59 | |||
JV048734.1 | - | 954 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |