Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257689.1 | NP_001244618.1 | 589 | 101 | ||
XM_028838539.1 | XP_028694372.1 | 727 | 101 | |||
GenBank mRNA | JU337184.1 | - | 589 | 101 | ||
JV633944.1 | - | 198 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |