Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257866.1 | NP_001244795.1 | 768 | 146 | ||
XM_015112950.2 | XP_014968436.1 | 772 | 146 | |||
XM_015112952.2 | XP_014968438.1 | 1279 | 146 | |||
XM_028831569.1 | XP_028687402.1 | 1315 | 146 | |||
XM_028831570.1 | XP_028687403.1 | 1234 | 146 | |||
GenBank mRNA | JU321483.1 | - | 768 | 146 | ||
JU472736.1 | - | 710 | 146 | |||
JV048764.1 | - | 633 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |