Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194217.2 | NP_001181146.1 | 558 | 135 | ||
GenBank mRNA | JU320692.1 | - | 566 | 135 | ||
JU473285.1 | - | 783 | 135 | |||
JU473286.1 | - | 651 | 135 | |||
JV048754.1 | - | 583 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |