Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258043.1 | NP_001244972.1 | 297 | 81 | ||
GenBank mRNA | JU470662.1 | - | 297 | 81 | ||
JU470663.1 | - | 297 | 81 | |||
JU470664.1 | - | 297 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |