Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194215.3 | NP_001181144.2 | 786 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |