Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261015.1 | NP_001247944.1 | 270 | 138 | ||
XM_015146873.2 | XP_015002359.1 | 306 | 138 | |||
XM_015146877.1 | XP_015002363.1 | 346 | 138 | |||
XM_015146878.2 | XP_015002364.1 | 354 | 138 | |||
XM_015146879.1 | XP_015002365.1 | 307 | 138 | |||
XM_028853537.1 | XP_028709370.1 | 301 | 133 | |||
GenBank mRNA | JU320819.1 | - | 370 | 138 | ||
JU485534.1 | - | 260 | 138 | |||
JV046919.1 | - | 270 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |