Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094268.4 | XP_001094268.2 | 918 | 80 | ||
GenBank mRNA | JV704019.1 | - | 843 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |