Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261140.1 | NP_001248069.1 | 468 | 107 | ||
GenBank mRNA | JU336535.1 | - | 468 | 107 | ||
JV594865.1 | - | 496 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |