Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107318.4 | XP_001107318.2 | 5867 | 80 | ||
XM_015150494.2 | XP_015005980.1 | 5868 | 80 | |||
XM_015150495.2 | XP_015005981.1 | 5868 | 80 | |||
GenBank mRNA | JU336120.1 | - | 5793 | 80 | ||
JU476982.1 | - | 5790 | 80 | |||
JU476983.1 | - | 5790 | 80 | |||
JV048076.1 | - | 5799 | 80 | |||
JV636342.1 | - | 5789 | 80 | |||
JV722099.1 | - | 5849 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |