Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261300.1 | NP_001248229.1 | 830 | 116 | ||
XM_028845416.1 | XP_028701249.1 | 407 | 116 | |||
GenBank mRNA | JU332453.1 | - | 923 | 116 | ||
JU473920.1 | - | 924 | 116 | |||
JV045061.1 | - | 830 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |