Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119349.2 | XP_014974835.2 | 1132 | 73 | ||
GenBank mRNA | JU323336.1 | - | 1089 | 73 | ||
JU473924.1 | - | 1103 | 73 | |||
JV045360.1 | - | 1073 | 73 |