Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266138.1 | NP_001253067.1 | 142 | 149 | ||
GenBank mRNA | JV045493.1 | - | 142 | 149 | ||
JV617375.1 | - | 278 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |