Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260635.1 | NP_001247564.1 | 757 | 112 | ||
XM_028846048.1 | XP_028701881.1 | 1053 | 112 | |||
GenBank mRNA | JU335335.1 | - | 729 | 112 | ||
JU476288.1 | - | 717 | 112 | |||
JV047569.1 | - | 757 | 112 | |||
JV550117.1 | - | 508 | 112 | |||
JV655457.1 | - | 765 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |