Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257444.1 | NP_001244373.1 | 929 | 107 | ||
XM_015127470.1 | XP_014982956.1 | 934 | 107 | |||
GenBank mRNA | JU322159.1 | - | 845 | 107 | ||
JU322225.1 | - | 929 | 107 | |||
JU471727.1 | - | 840 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |