Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265927.1 | NP_001252856.1 | 691 | 128 | ||
GenBank mRNA | JU320694.1 | - | 704 | 128 | ||
JU472808.1 | - | 680 | 128 | |||
JV045890.1 | - | 691 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |