Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113359.1 | NP_001106830.1 | 426 | 168 | ||
XM_015115035.2 | XP_014970521.1 | 739 | 168 | |||
GenBank mRNA | EU124697.1 | - | 426 | 168 | ||
JU332480.1 | - | 571 | 168 | |||
JU471017.1 | - | 577 | 168 | |||
JV045892.1 | - | 589 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |