Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099563.4 | XP_001099563.1 | 905 | 138 | ||
XM_001099664.4 | XP_001099664.1 | 1060 | 138 | |||
XM_001099759.4 | XP_001099759.1 | 776 | 138 | |||
XM_028851785.1 | XP_028707618.1 | 2057 | 138 | |||
XM_028851786.1 | XP_028707619.1 | 1967 | 138 | |||
XM_028851787.1 | XP_028707620.1 | 1589 | 138 | |||
XM_028851788.1 | XP_028707621.1 | 1364 | 138 | |||
XM_028851789.1 | XP_028707622.1 | 1720 | 138 | |||
XM_028851790.1 | XP_028707623.1 | 1630 | 138 | |||
XM_028851791.1 | XP_028707624.1 | 1567 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |