Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260854.1 | NP_001247783.1 | 3299 | 135 | ||
XM_028830422.1 | XP_028686255.1 | 2877 | 135 | |||
GenBank mRNA | JU321317.1 | - | 3072 | 135 | ||
JU321318.1 | - | 3072 | 135 | |||
JU476025.1 | - | 3299 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |