Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266627.1 | NP_001253556.1 | 643 | 147 | ||
XM_015123572.2 | XP_014979058.1 | 319 | 147 | |||
GenBank mRNA | JV043228.1 | - | 643 | 147 |
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU319753.1 | - | 245 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |