Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260534.1 | NP_001247463.1 | 842 | 80 | ||
XM_015144314.2 | XP_014999800.1 | 728 | 80 | |||
GenBank mRNA | JV045929.1 | - | 842 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |