Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195660.1 | NP_001182589.1 | 450 | 110 | ||
GenBank mRNA | CB308818.1 | - | 453 | 110 | ||
CO644659.1 | - | 436 | 110 | |||
JU326829.1 | - | 328 | 110 | |||
JU472778.1 | - | 438 | 110 | |||
JV045909.1 | - | 454 | 110 | |||
JV635773.1 | - | 462 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |