Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257882.1 | NP_001244811.1 | 279 | 78 | ||
XM_015142817.2 | XP_014998303.1 | 317 | 78 | |||
XM_015142818.2 | XP_014998304.1 | 192 | 78 | |||
GenBank mRNA | JU332526.1 | - | 279 | 78 | ||
JU474000.1 | - | 267 | 78 |