Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109351.4 | XP_001109351.2 | 3829 | 79 | ||
GenBank mRNA | JU325654.1 | - | 367 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |