Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113075.2 | XP_014968561.1 | 991 | 88 | ||
XM_028832278.1 | XP_028688111.1 | 991 | 88 | |||
XM_028832280.1 | XP_028688113.1 | 1261 | 88 | |||
GenBank mRNA | JU341876.1 | - | 742 | 88 | ||
JV648453.1 | - | 782 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |