Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260887.1 | NP_001247816.1 | 143 | 121 | ||
GenBank mRNA | JU332619.1 | - | 144 | 121 | ||
JU474082.1 | - | 157 | 121 | |||
JV045243.1 | - | 143 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |