Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266507.1 | NP_001253436.1 | 1422 | 125 | ||
XM_015152832.2 | XP_015008318.1 | 977 | 125 | |||
XM_015152833.2 | XP_015008319.1 | 1192 | 125 | |||
XM_028828950.1 | XP_028684783.1 | 1028 | 125 | |||
GenBank mRNA | JU323272.1 | - | 1422 | 125 | ||
JU472769.1 | - | 1422 | 125 | |||
JV044698.1 | - | 891 | 125 | |||
JV638460.1 | - | 933 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |