Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116124.2 | XP_014971610.2 | 2456 | 111 | ||
GenBank mRNA | JV569756.1 | - | 1510 | 111 | ||
JV705819.1 | - | 1229 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |