Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260908.1 | NP_001247837.1 | 963 | 64 | ||
GenBank mRNA | JV043752.1 | - | 963 | 64 | ||
JV635471.1 | - | 1011 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |