Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265940.1 | NP_001252869.1 | 706 | 153 | ||
GenBank mRNA | JU322968.1 | - | 706 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |