Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842551.1 | XP_028698384.1 | 1336 | 110 | ||
XM_028842552.1 | XP_028698385.1 | 1396 | 110 | |||
XM_028842553.1 | XP_028698386.1 | 944 | 110 | |||
GenBank mRNA | JV635739.1 | - | 1622 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |