Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082734.4 | XP_001082734.2 | 1882 | 116 | ||
GenBank mRNA | JU483501.1 | - | 1431 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |