Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082734.4 | XP_001082734.2 | 1961 | 94 | ||
GenBank mRNA | JU483501.1 | - | 1510 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |