Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257903.2 | NP_001244832.1 | 829 | 97 | ||
XM_028843154.1 | XP_028698987.1 | 1676 | 97 | |||
XM_028843156.1 | XP_028698989.1 | 1580 | 97 | |||
GenBank mRNA | JU323034.1 | - | 1005 | 97 | ||
JU323035.1 | - | 833 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |