Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265874.1 | NP_001252803.1 | 793 | 93 | ||
XM_015139520.2 | XP_014995006.1 | 836 | 93 | |||
GenBank mRNA | JU333948.1 | - | 712 | 93 | ||
JU475132.1 | - | 591 | 93 | |||
JV046732.1 | - | 793 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |