Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089547.4 | XP_001089547.4 | 1569 | 63 | ||
XM_001089893.4 | XP_001089893.3 | 1569 | 63 | |||
XM_028852340.1 | XP_028708173.1 | 1569 | 63 | |||
XM_028852344.1 | XP_028708177.1 | 1569 | 63 | |||
GenBank mRNA | JU322802.1 | - | 266 | 63 | ||
JU470903.1 | - | 198 | 63 | |||
JV047115.1 | - | 329 | 63 | |||
JV565127.1 | - | 191 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |