Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257650.1 | NP_001244579.1 | 376 | 95 | ||
GenBank mRNA | JU333731.1 | - | 376 | 95 | ||
JV046610.1 | - | 334 | 95 | |||
JV462926.1 | - | 261 | 95 | |||
JV647038.1 | - | 376 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |