Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194838.2 | NP_001181767.1 | 575 | 140 | ||
XM_001097426.4 | XP_001097426.1 | 854 | 140 | |||
XM_001097525.3 | XP_001097525.1 | 719 | 140 | |||
XM_028840479.1 | XP_028696312.1 | 717 | 140 | |||
XM_028840480.1 | XP_028696313.1 | 798 | 140 | |||
XM_028840481.1 | XP_028696314.1 | 710 | 140 | |||
GenBank mRNA | JU321653.1 | - | 716 | 140 | ||
JU471490.1 | - | 702 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |