Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193922.1 | NP_001180851.1 | 541 | 110 | ||
GenBank mRNA | JU332275.1 | - | 349 | 110 | ||
JU471591.1 | - | 316 | 110 | |||
JV565479.1 | - | 357 | 110 | |||
JV614237.1 | - | 355 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |