Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089493.4 | XP_001089493.1 | 360 | 132 | ||
GenBank mRNA | JU323227.1 | - | 319 | 132 | ||
JV045225.1 | - | 352 | 132 | |||
JV715747.1 | - | 345 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |