Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194241.3 | NP_001181170.1 | 208 | 97 | ||
XM_015121717.2 | XP_014977203.1 | 294 | 97 | |||
GenBank mRNA | JU332668.1 | - | 264 | 97 | ||
JU474102.1 | - | 178 | 97 | |||
JV046030.1 | - | 269 | 97 | |||
JV604213.1 | - | 291 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |