Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032872.1 | NP_001028044.1 | 885 | 111 | ||
XM_015131494.2 | XP_014986980.1 | 1121 | 111 | |||
XM_015131495.2 | XP_014986981.1 | 1121 | 111 | |||
GenBank mRNA | AF416334.1 | - | 885 | 111 | ||
JU484479.1 | - | 829 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |